Learn More
Cayman Chemical TrImetazIdInd8hydrochLRI 10mg
SDP
Catalog No. 502943187
Encompass_Preferred
An internal standard for the quantification of trimetazidine by GC- or LC-MS
Catalog No. 50-294-3187 Supplier Cayman Chemical Supplier No. 3981010 MG
An internal standard for the quantification of trimetazidine by GC- or LC-MS