Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimetazidine | 5011-34-7 | 99.8% | 266.34 g/mol | C14H22N2O3 | 10 MG
SDP

Catalog No. 5000296903
Encompass_Preferred

Trimetazidine is a small-molecule antianginal and cytoprotective research compound that modulates fatty acid metabolism and autophagy. It inhibits long-chain 3-ketoyl coenzyme A thiolase, reduces β-oxidation of free fatty acids, and has reported antioxidant and anti-inflammatory activities. Intended for laboratory research use only; not for human or clinical use.

  • Research-grade small molecule for laboratory studies.
  • Inhibits long-chain 3-ketoyl CoA thiolase and modulates fatty acid metabolism.
  • Exhibits antioxidant, anti-inflammatory, and cytoprotective properties.
  • Molecular formula C14H22N2O3; molecular weight 266.34 g/mol.
  • Supplied as a 10 MG quantity suitable for experimental dosing and formulation.

Catalog No. 50-002-96903 Supplier Medchemexpress LLC Supplier No. HYB0968A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trimetazidine is a small-molecule antianginal and cytoprotective research compound that modulates fatty acid metabolism and autophagy. It inhibits long-chain 3-ketoyl coenzyme A thiolase, reduces β-oxidation of free fatty acids, and has reported antioxidant and anti-inflammatory activities. Intended for laboratory research use only; not for human or clinical use.

  • Research-grade small molecule for laboratory studies.
  • Inhibits long-chain 3-ketoyl CoA thiolase and modulates fatty acid metabolism.
  • Exhibits antioxidant, anti-inflammatory, and cytoprotective properties.
  • Molecular formula C14H22N2O3; molecular weight 266.34 g/mol.
  • Supplied as a 10 MG quantity suitable for experimental dosing and formulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.