Learn More
Medchemexpress LLC Trimetazidine | 5011-34-7 | 99.84% | 50 MG
SDP
Catalog No. 5000409185
Encompass_Preferred
Trimetazidine is a selective long chain 3-ketoyl coenzyme A thiolase inhibitor, effective as an antianginal and cytoprotective agent. It possesses anti-oxidant, anti-inflammatory, antinociceptive, and gastroprotective properties, also triggering autophagy.
  • Inhibits β-oxidation of free fatty acid
  • Acts as an antianginal agent
  • Provides cytoprotective properties
  • Exhibits anti-oxidant properties
  • Offers anti-inflammatory properties
  • Has antinociceptive properties
  • Possesses gastroprotective properties
  • Triggers autophagy
  • Functions as a 3-hydroxyacyl-CoA dehydrogenase (HADHA) inhibitor
Catalog No. 50-004-09185 Supplier Medchemexpress LLC Supplier No. HYB0968A50MG
Trimetazidine is a selective long chain 3-ketoyl coenzyme A thiolase inhibitor, effective as an antianginal and cytoprotective agent. It possesses anti-oxidant, anti-inflammatory, antinociceptive, and gastroprotective properties, also triggering autophagy.
  • Inhibits β-oxidation of free fatty acid
  • Acts as an antianginal agent
  • Provides cytoprotective properties
  • Exhibits anti-oxidant properties
  • Offers anti-inflammatory properties
  • Has antinociceptive properties
  • Possesses gastroprotective properties
  • Triggers autophagy
  • Functions as a 3-hydroxyacyl-CoA dehydrogenase (HADHA) inhibitor