Learn More
Medchemexpress LLC HY-B0968 10mg , Trimetazidine (dihydrochloride) CAS:13171-25-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB096810MG
Encompass_Preferred
HY-B0968 10mg Trimetazidine (dihydrochloride) CAS: 13171-25-0 Trimetazidine dihydrochloride is a selective long chain 3-ketoyl coenzyme A thiolase inhibitor with an IC50 of 75 nM, which can inhibit β-oxidation of free fatty acid (FFA). Trimetazidine dihydrochloride is an effective antianginal agent and a cytoprotective drug, has anti-oxidant, anti-inflammatory, antinociceptive and gastroprotective properties. Trimetazidine dihydrochloride triggers autophagy. Trimetazidine dihydrochloride is also a 3-hydroxyacyl-CoA dehydrogenase (HADHA) inhibitor[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1927