Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimetazidine dihydrochloride | 13171-25-0 | MFCD00243086 | 99.9% | 339.26 | C14H24Cl2N2O3 | 10 MG
SDP

Catalog No. 5000297035
Encompass_Preferred

Trimetazidine dihydrochloride is the dihydrochloride salt of trimetazidine, used as an antianginal agent and as a reference standard in analytical and pharmacological research. Supplied in precisely weighed small pack sizes for assay and QC applications.

  • Reference standard for analytical and pharmacological assays.
  • Cas number 13171-25-0.
  • Chemical formula C14H24Cl2N2O3.
  • Molecular weight 339.26.
  • Purity 99.94% (as listed).
  • Available in small pack sizes such as 10 mg.

Catalog No. 50-002-97035 Supplier Medchemexpress LLC Supplier No. HYB0968R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trimetazidine dihydrochloride is the dihydrochloride salt of trimetazidine, used as an antianginal agent and as a reference standard in analytical and pharmacological research. Supplied in precisely weighed small pack sizes for assay and QC applications.

  • Reference standard for analytical and pharmacological assays.
  • Cas number 13171-25-0.
  • Chemical formula C14H24Cl2N2O3.
  • Molecular weight 339.26.
  • Purity 99.94% (as listed).
  • Available in small pack sizes such as 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.