Learn More
Selleck Chemical LLC Trimetazidine dihydrochloride S4543-500mg
SDP
Catalog No. 503110873
Encompass_Preferred
Trimetazidine dihydrochloride (Yoshimilon Kyurinett Vastarel F) is a kind of drug for treatment of chronic ischemic disorders It improves myocardial glucose utilization through inhibition of fatty acid metabolism known as fatty acid oxidation inhibitor
Catalog No. 50-311-0873 Supplier Selleck Chemical LLC Supplier No. S4543500MG
Trimetazidine dihydrochloride (Yoshimilon Kyurinett Vastarel F) is a kind of drug for treatment of chronic ischemic disorders It improves myocardial glucose utilization through inhibition of fatty acid metabolism known as fatty acid oxidation inhibitor