Learn More
TARGETMOL CHEMICALS INC Trimetazidine dihydrochloride
SDP
Catalog No. 502880727
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Trimetazidine dihydrochloride (Vastarel F) can improve myocardial glucose utilization by inhibiting fatty acid metabolism. Trimetazidine is the first cytoprotective anti-ischemic agent, also used as a vasodilator in ischemic heart disease or angina. Purity 99.31%
Catalog No. 50-288-0727 Supplier TARGETMOL CHEMICALS INC Supplier No. T0988500MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Trimetazidine dihydrochloride (Vastarel F) can improve myocardial glucose utilization by inhibiting fatty acid metabolism. Trimetazidine is the first cytoprotective anti-ischemic agent, also used as a vasodilator in ischemic heart disease or angina. Purity 99.31%