Learn More
Cayman Chemical TrImetazIdInhydrochlorIde 10mg
SDP
Catalog No. 502965003
Encompass_Preferred
An analytical reference standard categorized as a piperazine; has been detected as an AAF during anti-doping testing; intended for research and forensic applications
Catalog No. 50-296-5003 Supplier Cayman Chemical Supplier No. 1816510 MG
An analytical reference standard categorized as a piperazine; has been detected as an AAF during anti-doping testing; intended for research and forensic applications