Learn More
Cayman Chemical Trimetazidine (hydrochloride), Purity: ≥98%, CAS: 13171-25-0, 100 mg
SDP
Catalog No. 502965081
Encompass_Preferred
An analytical reference standard categorized as a piperazine; has been detected as an AAF during anti-doping testing; intended for research and forensic applications
Catalog No. 50-296-5081 Supplier Cayman Chemical Supplier No. 18165100 MG
An analytical reference standard categorized as a piperazine; has been detected as an AAF during anti-doping testing; intended for research and forensic applications