Learn More
Cayman Chemical Trimethadione, Purity: ≥98%, CAS: 127-48-0, 50 mg
SDP
Catalog No. 502994362
Encompass_Preferred
An anticonvulsant; decreases the levels of α-oxoglutarate, malate, and fumarate in the brain, as well as plasma glucose levels in rats; increases latency to first pentetrazole-induced clonic seizure in mice (ED50 = 10 mg/kg); inhibits absence-like seizures but not tonic convulsions in spontaneously epileptic rats
Catalog No. 50-299-4362 Supplier Cayman Chemical Supplier No. 2389350 MG
An anticonvulsant; decreases the levels of α-oxoglutarate, malate, and fumarate in the brain, as well as plasma glucose levels in rats; increases latency to first pentetrazole-induced clonic seizure in mice (ED50 = 10 mg/kg); inhibits absence-like seizures but not tonic convulsions in spontaneously epileptic rats