Learn More
Medchemexpress LLC HY-12751A 100mg Medchemexpress, Trimethobenzamide hydrochloride CAS:554-92-7 Purity:>98%
SDP
Catalog No. 502081133
Encompass_Preferred
Medchemexpress, HY-12751A 100mg Trimethobenzamide hydrochloride CAS:554-92-7 Trimethobenzamide hydrochloride is a blocker of the D2 receptor. Trimethobenzamide is an antiemetic used to prevent nausea and vomiting. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1133 Supplier Medchemexpress LLC Supplier No. HY12751A100MG
Medchemexpress, HY-12751A 100mg Trimethobenzamide hydrochloride CAS:554-92-7 Trimethobenzamide hydrochloride is a blocker of the D2 receptor. Trimethobenzamide is an antiemetic used to prevent nausea and vomiting. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.