Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethobenzamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T35251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35251G 1 g
T35255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35251G Supplier TCI America Supplier No. T35251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 554-92-7
Molecular Formula C21H29ClN2O5
Molecular Weight (g/mol) 424.922
MDL Number MFCD00057999
InChI Key WIIZEEPFHXAUND-UHFFFAOYSA-N
Synonym N-[4-[2-(Dimethylamino)ethoxy]benzyl]-3,4,5-trimethoxybenzamide Hydrochloride
PubChem CID 68385
IUPAC Name N-[[4-[2-(dimethylamino)ethoxy]phenyl]methyl]-3,4,5-trimethoxybenzamide;hydrochloride
SMILES CN(C)CCOC1=CC=C(C=C1)CNC(=O)C2=CC(=C(C(=C2)OC)OC)OC.Cl

Specifications

Melting Point 190°C
Color White
Quantity 1 g
Formula Weight 424.92
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethobenzamide Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.