Learn More
Cayman Chemical Trimethobenzamide (hydrochloride), Purity: ≥95%, CAS: 554-92-7, 50 mg
SDP
Catalog No. 502934660
Encompass_Preferred
A D2 receptor antagonist and antiemetic; prevents apomorphine-induced emesis in dogs when administered at a dose of 20 mg/kg; stimulates contraction of isolated guinea pig ileum (ED50 = 8.7 μg/mL); decreases apomorphine-induced pecking responses in pigeons by 20% and increases the latent period of pecking from 4.3 to 8 minutes when used at a dose of 20 mg/kg; enhances compensation and decreases recovery time from 70 to 32 weeks in unilaterally labyrinthectomized cats, a model of vestibular injury, when administered at a dose of 6.7 g/kg per day for 2 weeks
Catalog No. 50-293-4660 Supplier Cayman Chemical Supplier No. 2414550 MG
A D2 receptor antagonist and antiemetic; prevents apomorphine-induced emesis in dogs when administered at a dose of 20 mg/kg; stimulates contraction of isolated guinea pig ileum (ED50 = 8.7 μg/mL); decreases apomorphine-induced pecking responses in pigeons by 20% and increases the latent period of pecking from 4.3 to 8 minutes when used at a dose of 20 mg/kg; enhances compensation and decreases recovery time from 70 to 32 weeks in unilaterally labyrinthectomized cats, a model of vestibular injury, when administered at a dose of 6.7 g/kg per day for 2 weeks