Learn More
Cayman Chemical Trimethoprim, Purity: ≥98%, CAS: 738-70-5, 10 g
SDP
Catalog No. 502970518
Encompass_Preferred
A synthetic antibiotic that inhibits dihydrofolate reductase (DHFR), with prominent selectivity for bacterial DHFR over mammalian DHFR (IC50s = 5 and 30,000 nM, respectively)
Catalog No. 50-297-0518 Supplier Cayman Chemical Supplier No. 1647310 G
A synthetic antibiotic that inhibits dihydrofolate reductase (DHFR), with prominent selectivity for bacterial DHFR over mammalian DHFR (IC50s = 5 and 30,000 nM, respectively)