Learn More
Alkali Scientific Trimethoprim, 25 Grams
SDP
Supplier:  Alkali Scientific TRI003CP
Encompass_Preferred
Trimethoprim is an antibiotic used primarily in combination with other drugs to treat urinary tract infections, respiratory infections, and other bacterial diseases. This 25-gram powder form ensures a high concentration of the active ingredient, which works by inhibiting bacterial folic acid synthesis, a critical component of DNA replication. Trimethoprim is often paired with sulfamethoxazole to enhance its effectiveness and reduce bacterial resistance. The 25-gram format is typically used in hospital and clinical environments for compounding and formulation into suspension or other doses, offering efficient treatment for a wide range of bacterial infections.
Catalog No. 50-006-61054