Learn More
Research Products International Corp Trimethoprim, 25 Grams
SDP
Catalog No. 50213702
Encompass_Preferred
Trimethoprim is a bacteriostatic antibiotic that inhibits bacterial dihydrofolate reductase, an enzyme involved in the synthesis of tetrahydrofolic acid. This acid is crucial for the production of nucleic acids (DNA and RNA) in bacteria. By blocking this enzyme, trimethoprim disrupts the folate synthesis pathway, leading to the inhibition of bacterial growth.
Trimethoprim is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its presence in the culture medium allows for the specific isolation of bacteria that lack resistance to trimethoprim.
Catalog No. 50-213-702 Supplier Research Products International Corp Supplier No. T5900025.0
Trimethoprim is a bacteriostatic antibiotic that inhibits bacterial dihydrofolate reductase, an enzyme involved in the synthesis of tetrahydrofolic acid. This acid is crucial for the production of nucleic acids (DNA and RNA) in bacteria. By blocking this enzyme, trimethoprim disrupts the folate synthesis pathway, leading to the inhibition of bacterial growth.
Trimethoprim is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its presence in the culture medium allows for the specific isolation of bacteria that lack resistance to trimethoprim.