Learn More
Selleck Chemical LLC Trimethoprim E4837-1G
SDP
Catalog No. 5000041943
Encompass_Preferred
Trimethoprim (lactate) is an antifolate antibacterial agent that inhibits bacterial dihydrofolate reductase (DHFR) an essential enzyme responsible for the production of tetrahydrofolic acid (THF) By blocking this enzyme trimethoprim prevents the synthesis of bacterial DNA hindering bacterial survival It is a standard antibiotic for treating urinary tract infections (UTIs)
Catalog No. 50-000-41943 Supplier Selleck Chemical LLC Supplier No. E48371G
Trimethoprim (lactate) is an antifolate antibacterial agent that inhibits bacterial dihydrofolate reductase (DHFR) an essential enzyme responsible for the production of tetrahydrofolic acid (THF) By blocking this enzyme trimethoprim prevents the synthesis of bacterial DNA hindering bacterial survival It is a standard antibiotic for treating urinary tract infections (UTIs)