Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoprim, 99.8%, MP Biomedicals™

Catalog No. ICN19552725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19552725 25 g
ICN19552701 1 g
ICN19552705 5 g
ICN19552780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19552725 Supplier MP Biomedicals, Inc Supplier No. 0219552725
Add to Cart
Add to Cart

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.323
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
Synonym 2,4-Diamino-5-[3,4,5-trimethoxy-benzyl]pyrimidine
PubChem CID 5578
ChEBI CHEBI:45924
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N

Specifications

Type Trimethoprim
Physical Form Powder
Color White
Quantity 25 g
Assay Percent Range 0.998
Formula Weight 290.3
Chemical Name or Material Trimethoprim
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.