Learn More
Medchemexpress LLC Trimethoprim-d3 | 1189923-38-3 | 98.1% | 5 MG
SDP
Catalog No. 5000408572
Encompass_Preferred
Trimethoprim-d3 is the deuterium labeled Trimethoprim. Trimethoprim is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. It is active against a wide range of Gram-positive and Gram-negative aerobic bacteria. Trimethoprim has the potential for treating urinary tract infections, Shigellosis, and Pneumocystis pneumonia. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08572 Supplier Medchemexpress LLC Supplier No. HYB0510S25MG
Trimethoprim-d3 is the deuterium labeled Trimethoprim. Trimethoprim is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. It is active against a wide range of Gram-positive and Gram-negative aerobic bacteria. Trimethoprim has the potential for treating urinary tract infections, Shigellosis, and Pneumocystis pneumonia. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS