Learn More
Medchemexpress LLC Trimethoprim-d3 | 1189923-38-3 | MFCD16039497 | 99.2% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0510S21MG
Encompass_Preferred
Trimethoprim-d3 is the deuterium-labeled form of Trimethoprim, a bacteriostatic antibiotic that functions as an orally active dihydrofolate reductase inhibitor. It exhibits activity against a broad spectrum of Gram-positive and Gram-negative aerobic bacteria.
  • It can be utilized as a tracer in various applications.
  • It serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-09351