Learn More
Cayman Chemical TrImethoprIm-d3 500ug
SDP
Catalog No. 502934342
Encompass_Preferred
An internal standard for the quantification of trimethoprim by GC- or LC-MS
Catalog No. 50-293-4342 Supplier Cayman Chemical Supplier No. 26119500 UG
An internal standard for the quantification of trimethoprim by GC- or LC-MS