Learn More
Medchemexpress LLC Trimethoprim-d9 | 1189460-62-5 | 99.6% | 299.37 | C14H9D9N4O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0510S10MG
Encompass_Preferred
Trimethoprim-d9 is a deuterium-labeled analogue of the antibiotic trimethoprim, provided as a research-grade stable isotope standard for analytical and tracer applications. It is intended for laboratory research use only and not for human or veterinary use.
  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High chemical purity (99.6%).
  • Molecular formula C14H9D9N4O3; molecular weight 299.37 g/mol.
  • Supplied in a 10 MG quantity (other pack sizes available).
  • Suitable for quantitative analysis and method validation.
Catalog No. 50-002-98001