Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoprim, G-Biosciences
SCI_ED

Catalog No. S139833
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139833 100 g
S139814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139833 Supplier G-Biosciences Supplier No. RC1124
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trimethoprim is a bactericidal antibiotic effective against bacteria responsible for urinary tract infections, prostatitis, and cystitis.

It is commonly combined with sulfamethoxazole to combat Staphylococcus aureus.

Chemical Identifiers

Molecular Weight (g/mol) 290.32

Specifications

Physical Form Powder
Quantity 100 g
Product Line Gibco
Boiling Point 526.0°C
Percent Purity >98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.