Learn More
Medchemexpress LLC Trimethoprim hydrochloride | 60834-30-2 | 99.16% | 10 MM * 1 ML
SDP
Catalog No. 5000406977
Encompass_Preferred
Trimethoprim hydrochloride is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor, active against a wide range of Gram-positive and Gram-negative aerobic bacteria.
  • Potential for research into urinary tract infections, Shigellosis, and *Pneumocystis* pneumonia.
  • Inhibits infection of Influenza A virus in chick embryos when combined with zinc.
  • Bacteriostatic antibiotic.
  • Orally active dihydrofolate reductase inhibitor.
Catalog No. 50-004-06977 Supplier Medchemexpress LLC Supplier No. HYB0510B10MM/1ML
Trimethoprim hydrochloride is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor, active against a wide range of Gram-positive and Gram-negative aerobic bacteria.
  • Potential for research into urinary tract infections, Shigellosis, and *Pneumocystis* pneumonia.
  • Inhibits infection of Influenza A virus in chick embryos when combined with zinc.
  • Bacteriostatic antibiotic.
  • Orally active dihydrofolate reductase inhibitor.