Learn More
Medchemexpress LLC Trimethoprim hydrochloride | 60834-30-2 | 99.16% | 100 MG
SDP
Catalog No. 5000407597
Encompass_Preferred
Trimethoprim hydrochloride is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. It is active against a wide range of Gram-positive and Gram-negative aerobic bacteria. It has potential for research on urinary tract infections, Shigellosis, and Pneumocystis pneumonia. Trimethoprim hydrochloride can inhibit infection of Influenza A virus in chick embryo when combined with zinc.
  • Bacteriostatic antibiotic.
  • Orally active dihydrofolate reductase inhibitor.
  • Active against a wide range of Gram-positive and Gram-negative aerobic bacteria.
  • Potential for research of urinary tract infections, Shigellosis, and Pneumocystis pneumonia.
  • Can inhibit infection of Influenza A virus in chick embryo when combined with zinc.
Catalog No. 50-004-07597 Supplier Medchemexpress LLC Supplier No. HYB0510B100MG
Trimethoprim hydrochloride is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. It is active against a wide range of Gram-positive and Gram-negative aerobic bacteria. It has potential for research on urinary tract infections, Shigellosis, and Pneumocystis pneumonia. Trimethoprim hydrochloride can inhibit infection of Influenza A virus in chick embryo when combined with zinc.
  • Bacteriostatic antibiotic.
  • Orally active dihydrofolate reductase inhibitor.
  • Active against a wide range of Gram-positive and Gram-negative aerobic bacteria.
  • Potential for research of urinary tract infections, Shigellosis, and Pneumocystis pneumonia.
  • Can inhibit infection of Influenza A virus in chick embryo when combined with zinc.