Learn More
Medchemexpress LLC Trimethoprim hydrochloride | 60834-30-2 | 99.2% | 10 MG
SDP
Catalog No. 5000298916
Encompass_Preferred
Trimethoprim hydrochloride is a bacteriostatic antibiotic and a dihydrofolate reductase inhibitor supplied as the hydrochloride salt for research use in studies of bacterial infections and related biochemical assays.
  • high purity suitable for research use (HPLC 99.16%).
  • hydrochloride salt form improves solubility for solution preparation.
  • useful as a dihydrofolate reductase inhibitor in microbiology and pharmacology studies.
  • store sealed and protected from moisture; short-term storage at 4°C, in solvent: -20°C (1 month) or -80°C (6 months).
Catalog No. 50-002-98916 Supplier Medchemexpress LLC Supplier No. HYB0510B10MG
Trimethoprim hydrochloride is a bacteriostatic antibiotic and a dihydrofolate reductase inhibitor supplied as the hydrochloride salt for research use in studies of bacterial infections and related biochemical assays.
  • high purity suitable for research use (HPLC 99.16%).
  • hydrochloride salt form improves solubility for solution preparation.
  • useful as a dihydrofolate reductase inhibitor in microbiology and pharmacology studies.
  • store sealed and protected from moisture; short-term storage at 4°C, in solvent: -20°C (1 month) or -80°C (6 months).