Learn More
Medchemexpress LLC Trimethoprim hydrochloride | 60834-30-2 | >99.2% | 5 MILLIGRAM
SDP
Supplier:  Medchemexpress LLC HYB0510B5MG
Encompass_Preferred
Trimethoprim hydrochloride is a bacteriostatic antibiotic and an orally active dihydrofolate reductase inhibitor. It is effective against a broad spectrum of Gram-positive and Gram-negative aerobic bacteria. This compound shows potential for research into urinary tract infections, Shigellosis, and Pneumocystis pneumonia, and can inhibit Influenza A virus infection in chick embryos when combined with zinc.
  • Bacteriostatic antibiotic
  • Orally active dihydrofolate reductase inhibitor
  • Active against a wide range of Gram-positive and Gram-negative aerobic bacteria
  • Interrupts folate metabolism by inhibiting dihydrofolate reductase (DHFR)
  • Potential for research into urinary tract infections, Shigellosis, and Pneumocystis pneumonia
  • Can inhibit Influenza A virus infection in chick embryos when combined with zinc
Catalog No. 50-004-05817