Learn More
Alkali Scientific Trimethoprim Lactate, 250 Milligrams
Supplier: Alkali Scientific RIM001CP
Trimethoprim Lactate, 250 mg, is an antibiotic formulation used to treat bacterial infections, particularly urinary tract infections, and respiratory tract infections. This powder form is easily reconstituted and administered as an oral suspension or intravenous injection. Trimethoprim works by inhibiting bacterial folic acid synthesis, a crucial step in DNA and RNA synthesis. It is frequently used in combination with sulfamethoxazole for treating a wide range of infections, including those caused by both gram-negative and gram-positive bacteria. The 250mg dosage is commonly used for outpatient treatment and for managing uncomplicated infections.