Learn More
Crescent Chemical Co Inc TRIMETHOPRIM RESEARCH GRADE
SDP
Catalog No. 50248218
Encompass_Preferred
Trimethoprim research grade, Ph. Eur. Antibacterial substance; activity in vitro (1). Simultaneous detection with sulfamethazine by HPLC (2). Inhibitor of the bacterial enzyme dihydrofolate reductase. Size - 5G Storage Conditions - +2 °C TO +8 °C /
Catalog No. 50-248-218 Supplier Crescent Chemical Co Inc Supplier No. SE37049.01
Trimethoprim research grade, Ph. Eur. Antibacterial substance; activity in vitro (1). Simultaneous detection with sulfamethazine by HPLC (2). Inhibitor of the bacterial enzyme dihydrofolate reductase. Size - 5G Storage Conditions - +2 °C TO +8 °C /