Learn More
TOKU-E Trimethoprim ReadyMade™ Solution | 738-70-5 | 25 mg/ML in DMSO | filter-sterile | 10 x 1 ML
Catalog No. NC2068797
Encompass_Preferred
Trimethoprim ReadyMade™ Solution | 738-70-5 | 25 mg/ML in DMSO | filter-sterile | 10 x 1 ML
Catalog No. NC2068797 Supplier TOKU-E Supplier No. T14410X1ML
Trimethoprim ReadyMade™ Solution | 738-70-5 | 25 mg/ML in DMSO | filter-sterile | 10 x 1 ML