Learn More
AdipoGen Trimethoprim sulfate
SDP
Catalog No. 502559983
Encompass_Preferred
Chemical. CAS 56585-33-2. Formula C28H38N8O10S. MW 678.24. Synthetic Antibacterial compound. Pyrimidine inhibitor of dihydrofolate reductase.
Catalog No. 50-255-9983 Supplier AdipoGen Supplier No. CDXT0146M025
Chemical. CAS 56585-33-2. Formula C28H38N8O10S. MW 678.24. Synthetic Antibacterial compound. Pyrimidine inhibitor of dihydrofolate reductase.