Learn More
AdipoGen Trimethoprim sulfate
SDP
Catalog No. 502559982
Encompass_Preferred
Chemical. CAS 56585-33-2. Formula C28H38N8O10S. MW 678.24. Synthetic Antibacterial compound. Pyrimidine inhibitor of dihydrofolate reductase.
Catalog No. 50-255-9982 Supplier AdipoGen Supplier No. CDXT0146M100
Chemical. CAS 56585-33-2. Formula C28H38N8O10S. MW 678.24. Synthetic Antibacterial compound. Pyrimidine inhibitor of dihydrofolate reductase.