Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoprim USP, Medisca™
GREENER_CHOICESDP

Catalog No. 18048137 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-137 100 g
18-048-033 25 g
18-048-034 1 kg
18-048-092 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-048-137 Supplier Medisca Supplier No. 077005
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 738-70-5

Trimethoprim USP can be white to cream-colored, odorless crystals or crystalline powder. Particles micronized so that 90% are less than 20μm.

Chemical Identifiers

CAS 738-70-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline
Color Cream to White
Quantity 100 g
Percent Purity 98.5 to 101.0%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.