Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethoxy(2-phenylethyl)silane

Catalog No. 4383401G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-834-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-834-01G Supplier Sigma Aldrich Supplier No. 4383401G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethoxy(2-phenylethyl)silane (TMPES) is a phenyl functionalized trialkoxy silane that can be used as a siloxane based precursor.TMPES can be used as a silane-based coating on quantum dot (QD) light emitting diodes to improve the adhesion of QDs and improve the thermal stability of the device. It may be used in the synthesis of pure silica zeolite molecular sieves.

Specifications

CAS 49539-88-0
Density 1.033 g/mL (at 25°C)
Boiling Point 95°C to 96°C (2 mmHg)
Molecular Formula C11H18O3Si
Refractive Index n20/D 1.475 (literature)
Linear Formula C6H5CH2CH2Si(OCH3)3
Quantity 1 g
MDL Number MFCD00053852
Synonym (2-Phenylethyl)trimethoxysilane
Molecular Weight (g/mol) 226.34
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.