Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethoxy(3,3,3-Trifluoropropyl)Silane

Catalog No. 9187725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-877-25ML 25 mL
91-877-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 91-877-25ML Supplier Sigma Aldrich Supplier No. 9187725ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethoxy(3,3,3-trifluoropropyl)silane is commonly used in the synthesis of a variety of silsesquioxane based polymers. It is a precursor in the synthesis of macroporous gels for separation of oil and water. It can also be used in the synthesis of hydrophobic silica aerogel for the removal of uranium from aqueous solutions.

Specifications

CAS 429-60-7
Density 1.142 g/mL at 20°C
Boiling Point 144°C
Molecular Formula C6H13F3O3Si
Linear Formula C6H13F3O3Si
Refractive Index n20/D 1.355
MDL Number MFCD00039266
Quantity 25 mL
Synonym 3,3,3-Trifluoropropyl-trimethoxysilane
Molecular Weight (g/mol) 218.25 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.