Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethoxy(3,3,3-Trifluoropropyl)Silane

Catalog No. 918775ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-877-5ML 5 mL
91-877-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 91-877-5ML Supplier Sigma Aldrich Supplier No. 918775ML
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethoxy(3,3,3-trifluoropropyl)silane is commonly used in the synthesis of a variety of silsesquioxane based polymers. It is a precursor in the synthesis of macroporous gels for separation of oil and water. It can also be used in the synthesis of hydrophobic silica aerogel for the removal of uranium from aqueous solutions.

Specifications

CAS 126752-51-0
Molecular Formula Na4O32W10
Linear Formula Na4O32W10
Refractive Index n20/D 1.355
Quantity 5 mL
Synonym Sodium decatungstate
Molecular Weight (g/mol) 2442.34 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.