Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethoxy[3-(methylamino)propyl]silane

Catalog No. 551635100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-163-5100ML 100 mL
55-163-525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-163-5100ML Supplier Sigma Aldrich Supplier No. 551635100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethoxy[3-(methylamino)propyl]silane (MAPTMS) is an aminoalkyltrimethoxysilane that can be used for silanization surface immobilization.Trimethoxy[3-(methylamino)propyl]silane was coated onto a substrate for the preparation of gold dimers by aggregating gold nanospheres. The silane may be used as a silane coupling agent to prepare silica nanoparticles. 2 It may be used to modify quartz substrates.

Specifications

CAS 3069-25-8
Density 0.975 g/mL (at 25°C)
Boiling Point 106°C
Molecular Formula C7H19NO3Si
Linear Formula (CH3O)3 Si(CH2)3 NHCH3
Quantity 100 mL
MDL Number MFCD00048021
Synonym N-Methyl-3-(trimethoxysilyl)propylamine; [3-(Methylamino)propyl]trimethoxysilane
Molecular Weight (g/mol) 193.32
Percent Purity 95%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.