Learn More
Medchemexpress LLC Trimethoxy(4-phenylbutyl)silane | 152958-91-3 | 99.6% | 254.40 | 500 MG

Supplier: Medchemexpress LLC HY27980500MG
Trimethoxy(4-phenylbutyl)silane serves as a drug intermediate, crucial for the synthesis of various active compounds.
- Appearance: Liquid
- Color: Colorless to light yellow
- Chemical formula: C13H22O3Si
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.