Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethoxy(octyl)silane

Catalog No. 37622125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-622-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-622-125ML Supplier Sigma Aldrich Supplier No. 37622125ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethoxy(octyl)silane is an organosilane. The effect of hydrolyzed silane on the penetration of water into a lamellar liquid crystal was studied.Trimethoxy(octyl)silane was used for the silanization of silica nanoparticles. Surface modification of hydrophobic (T805) titania particles with the silane was reported.

Specifications

CAS 3069-40-7
Density 0.907 g/mL (at 25°C)
Molecular Formula C11H26O3Si
Refractive Index n20/D 1.416 (literature)
Linear Formula CH3(CH2)7 Si(OCH3)3
Quantity 25 mL
MDL Number MFCD00043053
Synonym Octyltrimethoxysilane
Molecular Weight (g/mol) 234.41
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.