Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoxy(pentafluorophenyl)silane 97.0+%, TCI America™
SDP

Catalog No. T33521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33521G 1 g
T33525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33521G Supplier TCI America Supplier No. T33521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 223668-64-2
Molecular Formula C9H9F5O3Si
Molecular Weight (g/mol) 288.245
MDL Number MFCD04039205
InChI Key XFFHTZIRHGKTBQ-UHFFFAOYSA-N
PubChem CID 2782640
IUPAC Name trimethoxy-(2,3,4,5,6-pentafluorophenyl)silane
SMILES CO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC

Specifications

Color Colorless
Quantity 1 g
Formula Weight 288.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(pentafluorophenyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.