Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethoxymethylsilane

Catalog No. 4401751L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-017-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 44-017-51L Supplier Sigma Aldrich Supplier No. 4401751L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethoxymethylsilane (MTMS) is one of the key precursor for the synthesis of:Monolithic silica columns with various skeleton sizes for capillary liquid chromatography.Ionogels, where an ionic liquid is confined within silica-derived networks.Monolithic silica aerogels via acid-base sol-gel polymerization.Hydrophobic, flexible, and ultralightweight silylated nanocellulose sponges for the selective removal of oil from water.

Specifications

CAS 89-79-2
Density 0.912 g/mL at 25°C
Boiling Point 212°C
Molecular Formula C10H18O
Refractive Index n20/D 1.371
Linear Formula C10H18O
MDL Number MFCD00134655
Quantity 1 L
Synonym MTMS; Methyltrimethoxysilane
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.