Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethoxymethylsilane

Catalog No. 246174250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-617-4250ML 250 mL
24-617-41L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 24-617-4250ML Supplier Sigma Aldrich Supplier No. 246174250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethoxymethylsilane (MTM) is an organosilicon compound widely used as a precursor for the preparation of silica-based materials, which finds the applications in various fields. Particularly in molecular assembly, linking nano building blocks, and selective synthesis oligosiloxane compounds. It can also be utilized as a crosslinker in the synthesis of polysiloxane polymers.Trimethoxymethylsilane can be used:As a silica source for synthesizing polyethyleneimine-silica (PEI-silica) organic-inorganic hybrid particles.To transform hydrophilic ceramic surfaces to hydrophobic by modifying the -OH groups.To modify silica aerogels by inducing hydrophobicity and enhancing mechanical properties without affecting transparency.

Specifications

CAS 1185-55-3
Density 0.955 g/mL at 25°C
Boiling Point 102°C to 104°C
Molecular Formula C4H12O3Si
Refractive Index n20/D 1.371
Linear Formula CH3Si(OCH3)3
MDL Number MFCD00008342
Quantity 250 mL
Synonym Methyltrimethoxysilane
Molecular Weight (g/mol) 136.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.