Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethoxyphenylsilane

Catalog No. 10474450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-474-450ML 50 mL
10-474-4250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-474-450ML Supplier Sigma Aldrich Supplier No. 10474450ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethoxyphenylsilane (TMPS) is an organotrialkoxysilane that is used as a functional monomer in the sol-gel processing.TMPS can also be used in the preparation of polytrimethoxyphenylsilane (PTMS) that facilitates the removal of graphene after chemical vapor deposition (CVD). Molecularly imprinted sol gels can be synthesized by using TMPS for the fabrication of localized surface plasmon resonance (LSPR).

Specifications

CAS 2996-92-1
Density 1.062 g/mL (at 25°C)
Boiling Point 233°C
Molecular Formula C9H14O3Si
Refractive Index n20/D 1.468 (literature)
Linear Formula C6H5Si(OCH3)3
Quantity 50 mL
MDL Number MFCD00025689
Synonym (Trimethoxysilyl)benzene; Phenyltrimethoxysilane
Molecular Weight (g/mol) 198.29
Percent Purity 97%
Biological Activity Bladder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.