Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoxysilane, 95%

Catalog No. AC392170050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392170050 5 g Glass bottle
AC392170250 25 g Glass bottle
AC392171000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC392170050 Supplier Thermo Scientific Chemicals Supplier No. 392170050
  • $79.00
    $56.65 / Each of 1

    Save $22.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2487-90-3 | C3H10O3Si | 122.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2487-90-3
Molecular Formula C3H10O3Si
Molecular Weight (g/mol) 122.2
MDL Number MFCD00008340
InChI Key PZJJKWKADRNWSW-UHFFFAOYSA-N
Synonym trimethoxysilane, silane, trimethoxy, trimethoxy silane, unii-v1j39xpf91, c3h10o3si, trimethoxysilyl, silanem trimethoxy, trimethoxysilyl radical, 4-01-00-01266 beilstein handbook reference
PubChem CID 6327428
IUPAC Name trimethoxysilicon
SMILES CO[Si](OC)OC

Specifications

Melting Point -115°C
Color Colorless
Density 0.9600g/mL
Boiling Point 86°C
Flash Point -9°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.3570 to 1.3600 (20°C, 589nm)
Linear Formula HSi(OCH3)3
Quantity 5 g
Specific Gravity 0.96
Solubility Information Solubility in water: reacts
Formula Weight 122.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Trimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Fatal if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moi
EINECSNumber 219-637-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.