Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl 1,3,5-Cyclohexanetricarboxylate (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. T30331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30331G 1 g
T30335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30331G Supplier TCI America Supplier No. T30331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52831-11-5
Molecular Formula C12H18O6
Molecular Weight (g/mol) 258.27
InChI Key ZZNOMLSSCRBRJS-UHFFFAOYSA-N
Synonym 1,3,5-Cyclohexanetricarboxylic Acid Trimethyl Ester
PubChem CID 247401
IUPAC Name trimethyl cyclohexane-1,3,5-tricarboxylate
SMILES COC(=O)C1CC(CC(C1)C(=O)OC)C(=O)OC

Specifications

Melting Point 46°C
Color White
Quantity 1 g
Formula Weight 258.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Trimethyl 1,3,5-Cyclohexanetricarboxylate (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.