Learn More
Medchemexpress LLC Trimethyl[2,3-(dioleyloxy)propyl]ammonium chloride | 104162-48-3 | MFCD00145456 | 98.0% | 670.57 g/mol | C42H84ClNO2 | 50 MG
SDP
Catalog No. 5000282662
Encompass_Preferred
Trimethyl[2,3-(dioleyloxy)propyl]ammonium chloride is a quaternary ammonium, lipid-like reagent used in life-science research. It acts as an amphiphilic cationic lipid for formulation and membrane studies, and can be used as a surfactant or helper lipid in experimental delivery systems. The material is supplied with product documentation and handling information and is intended for research use only.
  • Quaternary ammonium headgroup provides cationic charge.
  • Two dioleoyl chains confer lipid-like amphiphilicity.
  • Amphiphilic character supports bilayer and vesicle formulation.
  • Soluble in common organic solvents for formulation workflows.
  • Provided with documentation and safety data for lab use.
  • Store sealed and away from moisture; refrigerate for prolonged stability.
Catalog No. 50-002-82662 Supplier Medchemexpress LLC Supplier No. HYW12764450MG
Trimethyl[2,3-(dioleyloxy)propyl]ammonium chloride is a quaternary ammonium, lipid-like reagent used in life-science research. It acts as an amphiphilic cationic lipid for formulation and membrane studies, and can be used as a surfactant or helper lipid in experimental delivery systems. The material is supplied with product documentation and handling information and is intended for research use only.
  • Quaternary ammonium headgroup provides cationic charge.
  • Two dioleoyl chains confer lipid-like amphiphilicity.
  • Amphiphilic character supports bilayer and vesicle formulation.
  • Soluble in common organic solvents for formulation workflows.
  • Provided with documentation and safety data for lab use.
  • Store sealed and away from moisture; refrigerate for prolonged stability.