Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl[3-(trimethoxysilyl)propyl]ammonium Chloride (ca. 50% in Methanol), TCI America™
SDP

Catalog No. T279610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T279610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T279610G Supplier TCI America Supplier No. T279610G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35141-36-7
Molecular Formula C9H24ClNO3Si
Molecular Weight (g/mol) 257.83
MDL Number MFCD00054225
InChI Key FYZFRYWTMMVDLR-UHFFFAOYSA-M
Synonym n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium
PubChem CID 94462
IUPAC Name trimethyl(3-trimethoxysilylpropyl)azanium;chloride
SMILES C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-]

Specifications

Color Yellow
Quantity 10 g
UN Number 1230
Formula Weight 257.83
Physical Form Liquid
Chemical Name or Material Trimethyl[3-(trimethoxysilyl)propyl]ammonium Chloride (ca. 50% in Methanol)
EINECSNumber (2)-2094&(2)-2095
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.