Learn More
AdipoGen Trimethyl-4-sulfobutyl-indoliu
SDP
Catalog No. 502560036
Encompass_Preferred
Chemical. CAS 54136-26-4. Formula C15H21NO3S. MW 295.4. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.
Catalog No. 50-256-0036 Supplier AdipoGen Supplier No. CDXT0042M100
Chemical. CAS 54136-26-4. Formula C15H21NO3S. MW 295.4. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.