Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyl Borate

Catalog No. 44721825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-721-825ML 4 x 25 mL
44-721-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-721-825ML Supplier Sigma Aldrich Supplier No. 4472184X25ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethyl Borate

Specifications

CAS 121-43-7
Melting Point -34°C
Density 0.932 g/mL at 20°C
Boiling Point 68°C to 69°C
Molecular Formula C3H9BO3
Refractive Index n20/D 1.346
Linear Formula B(OCH3)3
MDL Number MFCD00008346
Quantity 4 x 25 mL
Synonym Boric acid trimethyl ester; Methyl borate
Molecular Weight (g/mol) 103.91 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.