Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyl Chitosan High Molecular Weight Degree Of Quaternization >70%

Catalog No. 9127001G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-270-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-270-01G Supplier Sigma Aldrich Supplier No. 9127001G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chitosan offers remarkable biological properties, it has been widely used in drug delivery and tissue engineering applications. Chitosan has good mucoadhesive properties due to its positive charge, which increases the adhesion to mucosa and facilitates drug penetration. Also, chitosan possesses hemostatic properties, which makes it a good candidate for wound dressing applications. The quaternization of the primary amine increases the water solubility of chitosan and keeps chitosan soluble over a wide pH range. Among all the quaternized chitosans, N, N, N-trimethyl chitosan chloride (TMC) is the most widely applied in biomedical applications.

Specifications

CAS 52349-26-5
Linear Formula (C9H18ClNO4)n
Quantity 1 g
Synonym N,N,N-Trimethyl chitosan; Chitosan trimethyl; Mucoadhesive polymers; TMC Chitosan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.